Tisk…
High visibility contrast traffic jacket, S466
Dodavatel: Portwest
S466RNRXL S466YERXS S466YER4XL S466YER5XL S466YERL S466YERM S466YERXL S466YERXXL S466YERXXXL S466YRBL S466YRBM S466YRBS S466YRBXL S466YRBXXL S466YRBXXXL S466YERS S466YBRXXXL S466YBRS S466YBRM S466YBRL S466YBRXL S466YBRXXL
PRTWS466RNRXLEA 1990 CZK
PRTWS466RNRXL PRTWS466YERXS PRTWS466YER4XL PRTWS466YER5XL PRTWS466YERL PRTWS466YERM PRTWS466YERXL PRTWS466YERXXL PRTWS466YERXXXL PRTWS466YRBL PRTWS466YRBM PRTWS466YRBS PRTWS466YRBXL PRTWS466YRBXXL PRTWS466YRBXXXL PRTWS466YERS PRTWS466YBRXXXL PRTWS466YBRS PRTWS466YBRM PRTWS466YBRL PRTWS466YBRXL PRTWS466YBRXXL
High visibility contrast traffic jacket, S466
Pracovní oděvy
Waterproof jacket made of 300D Oxford Weave polyester with PU coating. With nylon lining and wadding. Suitable for all weather conditions.
  • Two hip pockets with Velcro fastened flaps
  • Drawstring hood and hem
  • Knitted storm cuffs, taped waterproof seams
  • Two-way zip front with studded storm flap
  • Inside pocket, mobile phone pocket
Weight (outer fabric): 190 g/m²
Weight (polyester lining): 60 g/m²
Weight (polyester filling): 170 g/m²
Certifikace: EN ISO 20471 (Class 3), EN 343 (3-1), EN 342 (class 3)
Objednat
Produktová specifikace
Model S466
Environmentally Preferable
Aplikace oblečení Outdoor
Risk area Rain (waterproof)
Rod Unisex
Sleeve or leg length Long
Typ uzávěru Se zipem
Tkanina 300D Oxford Weave polyester with PU coating
Weight 420 g/m²