Tisk…
High visibility rain trousers, Flensburg 4448
Dodavatel: SIOEN
4448/FC1/1XL 4448/FC1/2XL 4448/FC1/3XL 4448/FC1/L 4448/FC1/M 4448/FC1/S
SIOE4448/FC1/1XLEA 2880 CZK
SIOE4448/FC1/1XL SIOE4448/FC1/2XL SIOE4448/FC1/3XL SIOE4448/FC1/L SIOE4448/FC1/M SIOE4448/FC1/S
High visibility rain trousers, Flensburg 4448
Pracovní oděvy
Waterproof and windproof trousers made from highly breathable Siopor® Extra fabric, consisting of 100% polyester fabric (cotton touch) with 100% PU film, with fixed polyamide lining.
  • Taped seams
  • Front fly with press studs
  • Two inset pockets
  • Legs adjustable by press studs, elasticated waist with drawstring
  • High tear resistance
The fabric is water-repellent on the outside and features a moisture attracting coating on the inside in order to keep the body warm and dry.
Weight: 205 g/m²
Certifikace: EN 471 (1-2), EN 343 (3-3)