Tisk…
Long swab rinse kits, SRK®
Dodavatel: Copan
902C 908C
710-0472EA 941 CZK
710-0472 710-0431
Long swab rinse kits, SRK®
Tampóny odběrové Tampóny s proplachovacím roztokem
Swab rinse kits (SRK®) for microbiological examination and quantitative analyses.
  • Swabs are separate from cap
  • Isotonic salt solution to neutralise and deactivate disinfectants and sanitising agents
  • Rinse solution used to moisten the sterile swab
Suitable for sampling and transporting the swab after collection.
Balení: Tube and applicator in plastic peel pouch - Each box contains 25 pouches.