Tisk…
Floor scales, Defender 3000, VE series
Dodavatel: OHAUS
80253278UK 80253279UK 80253275UK 80253276UK 80253277UK
611-3742EA 0 CZK
611-3742 611-3743 611-3739 611-3740 611-3741
Floor scales, Defender 3000, VE series
Váhy
Stainless steel platform or mild steel platform with 5 mm thick, fixed, top plate with four R60 alloy load cells with IP 67 rating. Easy to use balance with 5 weighing units and basic parts counting functionality.
  • T32XW indicator features stainless steel IP 65 housing with backlit LCD display
  • T31P indicator features a high impact ABS housing with keyholes on the rear cover for wall mounting
  • Internal universal power supply - rechargeable battery
  • IP 65 stainless steel junction box with side access
  • Easy to clean
  • Pit frames and ramps are available to fit your installation needs