Tisk…
Industrial gloves, Ultrane 548
Dodavatel: MAPA
34548080 34548087 34548088 34548086 34548089
MAPA34548080EA 828 CZK
MAPA34548080 MAPA34548087 MAPA34548088 MAPA34548086 MAPA34548089
Industrial gloves, Ultrane 548
Rukavice
These polyurethane foam gloves with seamless textile support and ventilated back provide light protection.
  • Optimal dexterity and sensitivity
  • Thin glove: excellent tactile sensitivity
  • Absorption of perspiration ensured by the cellular structure of the polyurethane
  • Particularly open texture of the specific lining helps improve hand ventilation
  • Anatomical shape: close fit for precision
Suitable for intricate assembly, sorting small parts, fitting small screws and fasteners, the assembly of electrical components, microprocessor handling and cosmetics preparation.
Certifikace: EN 388:2016 (3121X), Cat. II