Tisk…
Flame ionisation detector (FID) supplies
Dodavatel: Agilent
19231-60680 9301-0985 5188-5372 G3431-60680
HEWL19231-60680EA 2970 CZK
HEWL19231-60680 HEWL9301-0985 HEWL5188-5372 HEWLG3431-60680
Flame ionisation detector (FID) supplies
Chromatografické potřeby Chromatography Spare Parts
Before ordering parts for FID maintenance, determine which type of FID is installed on your GC. The FID is available in two versions: Dedicated, capillary optimized: for capillary columns only. Adaptable: for packed or capillary columns.
To determine the type of FID installed on your GC, open the oven door and examine the fitting at the base of the detector.