Tisk…

Thiosíran sodný 0.1 mol/l (0.1 N) in aqueous solution

Dodavatel: Lach-ner S.R.O.

Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate

62026010M1000 62026010L0010
LACH62026010M1000EA 280 CZK
LACH62026010M1000 LACH62026010L0010
Thiosíran sodný 0.1 mol/l (0.1 N) in aqueous solution
Thiosíran sodný
Vzorec: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS číslo: 7772-98-7
EINECS: 231-867-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR