Antimony(III) potassium oxitartrate trihydrate
Dodavatel: Lach-ner S.R.O.
|
Varování
|
Synonyms:
(R,R)-(+)-Antimony potassium tartrate trihydrate, (+)-Antimony potassium L-tartrate trihydrate, Tartar emetic, Antimony potassium L(+)-tartrate trihydrate, L(+)-Tartaric acid antimony potassium salt trihydrate, Potassium Antimony (III) Oxide Tartrate Trihydrate, (R,R)-(+)-Tartaric acid antimony potassium salt trihydrate, L(+)-Antimony potassium tartrate trihydrate, Potassium antimony(III) oxide tartrate trihydrate, Antimony potassium (+) tartrate trihydrate, Potassium antimony tartrate trihydrate, Potassium antimonyl tartrate sesquihydrate, Antimony potassium tartrate trihydrate, Potassium antimonyl tartrate trihydrate
Vzorec:
C₈H₄K₂O₁₂Sb₂·3H₂O MW: 667.88 Bod tání: 100 Hustota: 2.6 g/cm³ (20 °C) |
MDL Number:
MFCD00167056 CAS číslo: 28300-74-5 EINECS: 229-436-1 UN: 1551 ADR: 6.1 III Merck Index: 13,00706 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...