Tisk…

L(+)-vinan sodno-draselný tetrahydrát

Dodavatel: Lach-ner S.R.O.

Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate

40169CP0G1000 40169CP0G0500
LACH40169CP0G1000EA 790 CZK
LACH40169CP0G1000 LACH40169CP0G0500
L(+)-vinan sodno-draselný tetrahydrát
L(+)-vinan sodno-draselný tetrahydrát
Vzorec: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22
Bod tání: 70…80 °C
Hustota: ∼1.767 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS číslo: 6381-59-5
EINECS: 206-156-8
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR