Tisk…

Betaine 5 mol/l in aqueous solution, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: TMG

M-Clarity™ quality level = MQ200

B0300-1VL B0300-5VL
SIALB0300-1VLEA 832 CZK
SIALB0300-1VL SIALB0300-5VL
Betaine 5 mol/l in aqueous solution, Sigma-Aldrich®
Betaine
Vzorec: C₅H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00012123
CAS číslo: 107-43-7
EINECS: 203-490-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR