Tisk…
Baskets for Elmasonic S
Dodavatel: Elma
142-0013EA 5150 CZK
142-0013 142-0029 142-0044
Baskets for Elmasonic S
Koše a košíky Koše drátěné
These baskets feature a punched pattern on the base and side walls. In conjunction with a set of different pins, these holes allow the objects inside to be positioned as required. For unlimited flexibility. Each item can be individually secured – whether bulky industrial parts, laboratory glassware, small watch components or items of jewellery.
  • Made of stainless steel
  • With Rilsan-coated handles
  • Basket 200 also serves as a holder for sterilisation baskets
The number of objects that can be cleaned in the basket is thus maximised and optimal cleaning results are ensured thanks to perfect placement, which also saves water, cleaning chemicals, energy as well as time and money.