4-(Dimethylamino)benzaldehyd
Dodavatel: Lach-ner S.R.O.
Synonyms:
Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
40034AP0G0500
40034AP0G00501
40034AP0G0100
LACH40034AP0G0500EA
4420
CZK
LACH40034AP0G0500
LACH40034AP0G00501
LACH40034AP0G0100
4-(Dimethylamino)benzaldehyd
4-(Dimethylamino)benzaldehyde
Vzorec:
(CH₃)₂NC₆H₄CHO Bod varu: 176…177 °C (1013 hPa) Bod tání: 72 °C Hustota: 1,0254 g/cm³ (99.9 °C) Bod vzplanutí: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003381 CAS číslo: 100-10-7 EINECS: 202-819-0 Merck Index: 13,03257 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...