Dihydrogenfosforečnan sodný dihydrát
Dodavatel: Lach-ner S.R.O.
Synonyms:
Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate
30017AP0G0500
30017AP0G1000
LACH30017AP0G0500EA
405
CZK
LACH30017AP0G0500
LACH30017AP0G1000
Dihydrogenfosforečnan sodný dihydrát
Dihydrogenfosforečnan sodný dihydrát
Vzorec:
NaH₂PO₄·2H₂O MW: 156,01 g/mol Bod varu: ∼650 °C (1013 hPa) Bod tání: 60 °C Hustota: 1,91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149209 CAS číslo: 13472-35-0 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 12,08806 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...