Tisk…

Hydrogenfosforečnan dvojsodný dodekahydrát

Dodavatel: Lach-ner S.R.O.

Synonyms: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate

30061CP0G1000 30061-CP0-G 30061CP0G0500
LACH30061CP0G1000EA 474 CZK
LACH30061CP0G1000 LACH30061-CP0-G LACH30061CP0G0500
Hydrogenfosforečnan dvojsodný dodekahydrát
Hydrogenfosforečnan dvojsodný dodekahydrát
Vzorec: Na₂HPO₄·12H₂O
MW: 358.14
Bod tání: 35
Hustota: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149181
CAS číslo: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR