Molybdenan sodný dihydrát
Dodavatel: Lach-ner S.R.O.
Synonyms:
Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate
30127AP0G0500
30127AP0G1000
LACH30127AP0G0500EA
3330
CZK
LACH30127AP0G0500
LACH30127AP0G1000
Molybdenan sodný dihydrát
Molybdenan sodný dihydrát
Vzorec:
Na₂MoO₄·2H₂O MW: 241.95 Bod tání: 687 Hustota: 3.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149170 CAS číslo: 10102-40-6 EINECS: 231-551-7 Merck Index: 13,08718 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...