Methyl blue
Dodavatel: Lach-ner S.R.O.
Synonyms:
C.I. 42780
40101IP0G00101
40101IP0G00501
40101IP0G01001
40101IP0G0100
LACH40101IP0G00101EA
0
CZK
LACH40101IP0G00101
LACH40101IP0G00501
LACH40101IP0G01001
LACH40101IP0G0100
Methyl blue
Methyl blue
Vzorec:
C₃₇H₂₇N₃Na₂O₉S₃ Storage Temperature: Ambient |
MDL Number:
MFCD00058509 CAS číslo: 28983-56-4 EINECS: 249-352-9 Merck Index: 13,06055 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...