Kyselina 4-amino-3-hydroxy-1-naftalensulfonová
Dodavatel: Lach-ner S.R.O.
Synonyms:
1-Amino-2-naphthol-4-sulphonic acid, 4-Amino-3-hydroxy-α-naphthalenesulphonic acid, 4-Amino-3-hydroxynaphthalene-1-sulphonic acid, 1-Amino-2-hydroxy-4-naphthalenesulphonic acid
10008
10008AP0G00251
LACH10008EA
0
CZK
LACH10008
LACH10008AP0G00251
Kyselina 4-amino-3-hydroxy-1-naftalensulfonová
Kyselina 4-amino-3-hydroxy-1-naftalensulfonová
Vzorec:
C₁₀H₉NO₄S MW: 239.25 Bod varu: 466 Bod tání: ∼285 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004019 CAS číslo: 116-63-2 EINECS: 204-147-3 Merck Index: 13,00453 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...