Tisk…

Methylene blue trihydrate

Dodavatel: Lach-ner S.R.O.
Varování

Synonyms: 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride trihydrate, C.I.52015, Methylthioninium chloride trihydrate, Basic blue 9, Swiss Blue, Methylene blue

40100IP0G0100 40100IP0G1000 40100-IP0- 40100IP0G0025
LACH40100IP0G0100EA 2760 CZK
LACH40100IP0G0100 LACH40100IP0G1000 LACH40100-IP0- LACH40100IP0G0025
Methylene blue trihydrate
Methylene blue trihydrate
Vzorec: C₁₆H₁₈ClN₃S·3H₂O
MW: 373.9
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS číslo: 7220-79-3
EINECS: 200-515-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR