Fluorescein dvojsodná sůl
Dodavatel: Lach-ner S.R.O.
Synonyms:
3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate
40050-BP0-G
40050BP0G0100
40050BP0G0500
40050BP0G10001
LACH40050-BP0-GEA
0
CZK
LACH40050-BP0-G
LACH40050BP0G0100
LACH40050BP0G0500
LACH40050BP0G10001
Fluorescein dvojsodná sůl
Fluorescein dvojsodná sůl
Vzorec:
C₂₀H₁₀Na₂O₅ Bod tání: > 300 °C Hustota: 1,601 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00167039 CAS číslo: 518-47-8 EINECS: 208-253-0 Merck Index: 13,04185 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...