Fluorescein
Dodavatel: Lach-ner S.R.O.
Synonyms:
3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
40051-IP0-G
40051IP0G0100
LACH40051-IP0-GEA
317
CZK
LACH40051-IP0-G
LACH40051IP0G0100
Fluorescein
Fluorescein
Vzorec:
C₂₀H₁₂O₅ MW: 332.31 Bod tání: 314…316 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005050 CAS číslo: 2321-07-5 EINECS: 219-031-8 Merck Index: 13,04185 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...