Chlorid manganatý tetrahydrát
Dodavatel: Lach-ner S.R.O.
|
Varování
|
Synonyms:
Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate
30087AP0G05001
30087AP0G1000
LACH30087AP0G05001EA
831
CZK
LACH30087AP0G05001
LACH30087AP0G1000
Chlorid manganatý tetrahydrát
Chlorid manganatý tetrahydrát
Vzorec:
MnCl₂·4H₂O Bod varu: 1190 °C (1013 hPa) Bod tání: 58…59 °C Hustota: 2,01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149792 CAS číslo: 13446-34-9 EINECS: 231-869-6 UN: 3077 ADR: 9,III Merck Index: 13,05751 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...