Tisk…

Glycin

Dodavatel: Lach-ner S.R.O.

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

40059AP0G0500 40059AP0G1000
LACH40059AP0G0500EA 1990 CZK
LACH40059AP0G0500 LACH40059AP0G1000
Glycin
Glycin
Vzorec: NH₂CH₂COOH
MW: 75.07
Bod varu: 181
Bod tání: 83
Hustota: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS číslo: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR