Tisk…

Kyselina L(+)-glutamová

Dodavatel: Lach-ner S.R.O.

Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

10041AP0G0100 10041AP0G02501
LACH10041AP0G0100EA 0 CZK
LACH10041AP0G0100 LACH10041AP0G02501
Kyselina L(+)-glutamová
Kyselina L(+)-glutamová
Vzorec: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147,13 g/mol
Bod varu: 325 °C (1013 hPa)
Bod tání: 160 °C
Hustota: 1,538 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS číslo: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR