Hexakyanoželezitan draselný
Dodavatel: Lach-ner S.R.O.
Synonyms:
Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide
30053AP0G0500
30053AP0G1000
LACH30053AP0G0500EA
0
CZK
LACH30053AP0G0500
LACH30053AP0G1000
Hexakyanoželezitan draselný
Hexakyanoželezitan draselný
Vzorec:
K₃Fe(CN)₆ MW: 329,25 g/mol Hustota: 1,85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011392 CAS číslo: 13746-66-2 EINECS: 237-323-3 Merck Index: 13,07712 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...