thiokyanatan amonný
Dodavatel: Lach-ner S.R.O.
|
Varování
|
Synonyms:
Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate
30202AP0G05001
30202AP0G01001
30202AP0G1000
LACH30202AP0G05001EA
974
CZK
LACH30202AP0G05001
LACH30202AP0G01001
LACH30202AP0G1000
thiokyanatan amonný
thiokyanatan amonný
Vzorec:
NH₄SCN MW: 76.12 Bod tání: 150 Hustota: 1.305 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011428 CAS číslo: 1762-95-4 EINECS: 217-175-6 Merck Index: 13,00564 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...