Dihydrogenfosforečnan amonný
Dodavatel: Lach-ner S.R.O.
Synonyms:
Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate
30015AP0G0500
30015AP0G1000
LACH30015AP0G0500EA
507
CZK
LACH30015AP0G0500
LACH30015AP0G1000
Dihydrogenfosforečnan amonný
Dihydrogenfosforečnan amonný
Vzorec:
NH₄H₂PO₄ MW: 115.03 Bod tání: 190 Hustota: 1.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003396 CAS číslo: 7722-76-1 EINECS: 231-764-5 Merck Index: 13,00547 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...