Tisk…

Hydrogendifluorid amonný

Dodavatel: Lach-ner S.R.O.
Nebezpečí

Synonyms: Ammonium bifluoride, Ammonium hydrogen fluoride

30058CP0G1000 30058CP0G0500
LACH30058CP0G1000EA 0 CZK
LACH30058CP0G1000 LACH30058CP0G0500
Hydrogendifluorid amonný
Hydrogendifluorid amonný
Vzorec: NH₄HF₂
MW: 57,04 g/mol
Bod tání: 127 °C
Hustota: 0,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS číslo: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR