L(+)-Hydrogenvinan draselný
Dodavatel: Lach-ner S.R.O.
Synonyms:
Potassium hydrogen L-tartrate, Potassium acid tartrate, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Potassium Bitartrate, (R,R)-(+)-Tartaric acid potassium salt, L(+)-Tartaric acid potassium salt
40065AP0G100_S
40065AP0G0500
LACH40065AP0G100_SEA
0
CZK
LACH40065AP0G100_S
LACH40065AP0G0500
L(+)-Hydrogenvinan draselný
L(+)-Hydrogenvinan draselný
Vzorec:
KOOC(CHOH)₂COOH MW: 188.18 Bod tání: 250 Hustota: 1.943 g/cm³ (20 °C) |
MDL Number:
MFCD00065392 CAS číslo: 868-14-4 EINECS: 212-769-1 Merck Index: 13,07697 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...