4-Phenyl-1-butene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
P20602-10ML
P20602-50ML
SIALP20602-10MLEA
3960
CZK
SIALP20602-10ML
SIALP20602-50ML
4-Phenyl-1-butene, Sigma-Aldrich®
4-Phenyl-1-butene
Vzorec:
C₁₀H₁₂ |
MDL Number:
MFCD00026111 CAS číslo: 768-56-9 EINECS: 405-980-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...