Histamine (free base), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-(1H-Imidazol-4-yl)ethan-1-amine, Histamine
M-Clarity™ quality level = MQ200
H7125-1G
H7125-25G
H7125-5G
SIALH7125-1GEA
3290
CZK
SIALH7125-1G
SIALH7125-25G
SIALH7125-5G
Histamine (free base), Sigma-Aldrich®
Histamine (free base)
Vzorec:
C₅H₉N₃ MW: 111,15 g/mol |
MDL Number:
MFCD00005210 CAS číslo: 51-45-6 EINECS: 200-100-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...