Tisk…

Sunflower seed oil, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

S5007-1L S5007-250ML
SIALS5007-1LEA 8360 CZK
SIALS5007-1L SIALS5007-250ML
Sunflower seed oil, Sigma-Aldrich®
Sunflower seed oil
MDL Number: MFCD00132403
CAS číslo: 8001-21-6
EINECS: 232-273-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR