Tisk…

DL-3-Aminobutyric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Aminobutanoic acid

M-Clarity™ quality level = MQ100

A44207-25G A44207-5G
SIALA44207-25GEA 8290 CZK
SIALA44207-25G SIALA44207-5G
DL-3-Aminobutyric acid, Sigma-Aldrich®
DL-3-Aminobutyric acid
Vzorec: C₄H₉NO₂
MW: 103,1198 g/mol
MDL Number: MFCD00008087
CAS číslo: 541-48-0
EINECS: 208-783-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR