6-Methoxy-2-naphthaleneboronic acid 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
6-Methoxy-2-naphthylboronic acid, 6-Methoxy-2-naphthylboronic Acid
L19060.03
L19060.06
L19060.03EA
951
CZK
L19060.03
L19060.06
6-Methoxy-2-naphthaleneboronic acid 95%
6-Methoxy-2-naphthaleneboronic acid
Purity: 95%
CAS No.: 156641-98-4
Molecular Formula: C11H11BO3
Formula Weight: 202.02
Melting Point: ca 245° dec.
Beilstein Reference: 7636998
Vzorec:
C₁₁H₁₁BO₃ MW: 202,02 g/mol Bod tání: ∼ 245 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD03093087 CAS číslo: 156641-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...