4-Carboxy-1-naphthaleneboronic acid 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H56542.MD
H56542.03
H56542.MDEA
6040
CZK
H56542.MD
H56542.03
4-Carboxy-1-naphthaleneboronic acid 95%
4-Carboxy-1-naphthaleneboronic acid
Vzorec:
C₁₁H₉BO₄ Storage Temperature: Ambient |
MDL Number:
MFCD06203538 CAS číslo: 332398-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...