1-Fluoropyridiniumtrifluoromethanesulphonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
N-Fluoropyridinium triflate, 1-Fluoropyridiniumtriflate, N-Fluoropyridinium trifluoromethanesulphonate
M-Clarity™ quality level = MQ100
323659-1G
323659-5G
SIAL323659-1GEA
3130
CZK
SIAL323659-1G
SIAL323659-5G
1-Fluoropyridiniumtrifluoromethanesulphonate, Sigma-Aldrich®
1-Fluoropyridiniumtrifluoromethanesulphonate
Vzorec:
C₆H₅F₄NO₃S |
MDL Number:
MFCD00013458 CAS číslo: 107263-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...