Tisk…

Histamine dihydrochloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Histamine 2HCl, 4-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride

53300-1G 53300-25G 53300-5G
SIAL53300-1GEA 613 CZK
SIAL53300-1G SIAL53300-25G SIAL53300-5G
Histamine dihydrochloride, Sigma-Aldrich®
Histamine dihydrochloride
Vzorec: C₅H₁₁Cl₂N₃
MW: 184.07 g/mol
MDL Number: MFCD00012703
CAS číslo: 56-92-8
EINECS: 200-298-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR