Histamine dihydrochloride 99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Histamine 2HCl, 4-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride
150620050
150620250
150621000
150625000
ACRO150620050EA
3400
CZK
ACRO150620050
ACRO150620250
ACRO150621000
ACRO150625000
Histamine dihydrochloride 99%
Histamine dihydrochloride
Vzorec:
C₅H₁₁Cl₂N₃ MW: 184,07 g/mol Bod tání: 249…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012703 CAS číslo: 56-92-8 EINECS: 200-298-4 |
Výsledky specifikačního testu
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (105°C) |
Sulfated ash | ≤0.1 % |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...