Tisk…

2-Aminoimidazole hemisulphate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Aminoimidazole sulphate, 1H-imidazol-2-amine sulphate(2:1)

M-Clarity™ quality level = MQ100

197912-2.5G 197912-10G
SIAL197912-2.5GEA 2740 CZK
SIAL197912-2.5G SIAL197912-10G
2-Aminoimidazole hemisulphate, Sigma-Aldrich®
2-Aminoimidazole hemisulphate
Vzorec: C₆H₁₂N₆O₄S
MW: 264.26 g/mol
MDL Number: MFCD00013162
CAS číslo: 1450-93-7
EINECS: 215-918-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR