1-Methyl-2-vinylpyridinium trifluoromethanesulphonate, Sigma-Aldrich®
Dodavatel: Merck
69701-100MG
69701-500MG
SIAL69701-100MGEA
0
CZK
SIAL69701-100MG
SIAL69701-500MG
1-Methyl-2-vinylpyridinium trifluoromethanesulphonate, Sigma-Aldrich®
1-Methyl-2-vinylpyridinium trifluoromethanesulphonate
Vzorec:
C₉H₁₀F₃NO₃S |
MDL Number:
MFCD00210068 CAS číslo: 339530-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...