1-Vinylimidazole ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1-Vinyl-1H-imidazole
L16174.14
L16174.22
L16174.14EA
834
CZK
L16174.14
L16174.22
1-Vinylimidazole ≥99%
1-Vinylimidazole
Purity: 99%
CAS No.: 1072-63-5
Molecular Formula: C5H6N2
Formula Weight: 94.12
Flash Point: 84°(183°F)
Refractive Index: 1.5315
Melting Point:
Vzorec:
C₅H₆N₂ Bod varu: 192…194 °C Bod tání: < –50 °C Hustota: 1,038 Bod vzplanutí: 84 °C (183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005297 CAS číslo: 1072-63-5 EINECS: 214-012-0 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...