Tisk…

(Chloromethylene)dimethylammonium chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-Chloromethylen-N,N-Dimethyl Ammonium Chloride

M-Clarity™ quality level = MQ200

280909-25G 280909-5G
SIAL280909-25GEA 5020 CZK
SIAL280909-25G SIAL280909-5G
(Chloromethylene)dimethylammonium chloride, Sigma-Aldrich®
(Chloromethylene)dimethylammonium chloride
Vzorec: C₃H₇Cl₂N
MW: 128,00 g/mol
MDL Number: MFCD00011868
CAS číslo: 3724-43-4
EINECS: 425-970-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR