1-Pyrenebutyric acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A17760.06
A17760.03
A17760.06EA
5560
CZK
A17760.06
A17760.03
1-Pyrenebutyric acid ≥97%
1-Pyrenebutyric acid
Purity: 97%
CAS No.: 3443-45-6
Molecular Formula: C20H16O2
Formula Weight: 288.35
Melting Point: 184-186°
Beilstein Reference: 2140554
Vzorec:
C₂₀H₁₆O₂ Bod tání: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004141 CAS číslo: 3443-45-6 EINECS: 222-354-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...