Tisk…

Benzyl (S)-(+)-glycidyl ether ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (S)-2-((Benzyloxy)methyl)oxirane

L14033.03 L14033.06
L14033.03EA 1280 CZK
L14033.03 L14033.06
Benzyl (S)-(+)-glycidyl ether ≥98%
Benzyl (S)-(+)-glycidyl ether

Purity: 98+% CAS No.: 16495-13-9 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 110°(230°F) Refractive Index: 1.5170 Boiling Point: 130°/0.1mm Beilstein Reference: 5246495 Optical Rotation: +5.1° (c=5 in toluene) MDL No.: MFCD00054428

Vzorec: C₁₀H₁₂O₂
MW: 164,20 g/mol
Bod varu: 130 °C (0,1 mmHg)
Hustota: 1,072
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054428
CAS číslo: 16495-13-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR