(R)-(-)-Glycidyl methyl ether, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
454680-1G
454680-5G
SIAL454680-1GEA
2980
CZK
SIAL454680-1G
SIAL454680-5G
(R)-(-)-Glycidyl methyl ether, Sigma-Aldrich®
(R)-(-)-Glycidyl methyl ether
Vzorec:
C₄H₈O₂ |
MDL Number:
MFCD00273372 CAS číslo: 64491-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...