Tisk…

Serotonin hydrochloride

Dodavatel: Thermo Scientific

Synonyms: 5-Hydroxytryptamine Hydrochloride

S21263.A1 S21263.09
S21263.A1EA 0 CZK
S21263.A1 S21263.09
Serotonin hydrochloride
Serotonin hydrochloride
Vzorec: C₁₀H₁₃ClN₂O
MW: 212,68 g/mol
MDL Number: MFCD00012686
CAS číslo: 153-98-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR