Methanol
Dodavatel: Lach-ner S.R.O.
|
Nebezpečí
|
Synonyms:
Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
20038AT0M2500
20038AT0M5000
20038AT0G7900
20038AT0M1000
LACH20038AT0M2500EA
383
CZK
LACH20038AT0M2500
LACH20038AT0M5000
LACH20038AT0G7900
LACH20038AT0M1000
Methanol
Methanol
Vzorec:
CH₃OH MW: 32.04 Bod varu: 64.6 Bod tání: –98 °C Hustota: 0.7918 g/cm³ (20 °C) Bod vzplanutí: 11 °C (ot. pohárek) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS číslo: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3 II REACH: 01-2119433307-44 Merck Index: 13,05984 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...