Tisk…

Chloroform Stabilised, pure

Dodavatel: Lach-ner S.R.O.
Nebezpečí

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Amylen

20034CT1M1000 20034CT1M2500
LACH20034CT1M1000EA 419 CZK
LACH20034CT1M1000 LACH20034CT1M2500
Chloroform Stabilised, pure
Chloroform
Vzorec: CHCl₃
MW: 119.38
Bod varu: 61.7
Bod tání: –63 °C
Hustota: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS číslo: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR