Dichlormethan
Dodavatel: Lach-ner S.R.O.
|
Varování
|
Synonyms:
Methylene chloride
20020CT1M5000
20020-CT1-M
20020CT1M1000
LACH20020CT1M5000EA
1380
CZK
LACH20020CT1M5000
LACH20020-CT1-M
LACH20020CT1M1000
Dichlormethan
Dichloromethane
Vzorec:
CH₂Cl₂ MW: 84.93 Bod varu: 39.8 Bod tání: –95 °C Hustota: 1.322 Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS číslo: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1 III Merck Index: 12,06140 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...