Izoamylacetát 99% cont. ca 10% other isomers
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Isopentyl acetate, Acetic acid isoamyl ester
B21618.AE
B21618.AP
B21618.AEEA
824
CZK
B21618.AE
B21618.AP
Izoamylacetát 99% cont. ca 10% other isomers
Izoamylacetát
Purity: 99%
CAS No.: 123-92-2
Molecular Formula: CH3CO2CH2CH2CH(CH3)2
Formula Weight: 130.19
Flash Point: 25°(77°F)
Refractive Index: 1.400
Odour: Fruit-like
Melting Point: -78°
Boiling Point: 142°
Merck Reference: 14,5111
UN No.: 1104
MDL No.: MFCD00008946
Vzorec:
C₇H₁₄O₂ MW: 130,19 g/mol Bod varu: 142 °C Bod tání: –78 °C Hustota: 0,876 Bod vzplanutí: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008946 CAS číslo: 123-92-2 EINECS: 204-662-3 UN: 1104 ADR: 3,III Merck Index: 14,5111 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...