Acetonitril
Dodavatel: Lach-ner S.R.O.
|
Nebezpečí
|
Synonyms:
Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
20041AT0M5000
20041AT0M1000
LACH20041AT0M5000EA
2790
CZK
LACH20041AT0M5000
LACH20041AT0M1000
Acetonitril
Acetonitril
Vzorec:
H₃CCN MW: 41.05 Bod varu: 81.6 Bod tání: –45.7 °C Hustota: 0.782 g/cm³ (20 °C) Bod vzplanutí: 2 °C (ot. pohárek) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS číslo: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3 II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...